Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tiaramide Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. T3176100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T3176100MG 100 mg
T317625MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T3176100MG Supplier TCI America Supplier No. T3176100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 35941-71-0
Molecular Formula C15H21Cl2N3O3S
Molecular Weight (g/mol) 394.31
MDL Number MFCD01702832
InChI Key XJTXBQFPQSKSPV-UHFFFAOYNA-N
Synonym 5-Chloro-3-[[4-(2-hydroxyethyl)piperazin-1-yl]carbonylmethyl]-2-benzothiazolinone Hydrochloride
PubChem CID 443949
IUPAC Name hydrogen 5-chloro-3-{2-[4-(2-hydroxyethyl)piperazin-1-yl]-2-oxoethyl}-2,3,3a,7a-tetrahydro-1,3-benzothiazol-2-one chloride
SMILES [H+].[Cl-].OCCN1CCN(CC1)C(=O)CN1C2C=C(Cl)C=CC2SC1=O

Specifications

Melting Point 160°C
Color White-Yellow
Quantity 100 mg
Formula Weight 392.30
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tiaramide Hydrochloride
RTECSNumber TL7050000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.