Learn More
ATCC HEL 92.1.7 (ATCC TIB-180) Human Erythroleukemia Cell Line, Suspension Culture
Supplier:  ATCC HEL92.1.7
Encompass
HEL 92.1.7 (ATCC TIB-180) is a human erythroleukemia cell line derived from bone marrow and used for immunology and hematopoietic research. The line grows in suspension and can undergo erythroid or macrophage-like differentiation under specific treatments. Maintain cultures in RPMI-1640 with 10% fetal bovine serum at 37°C and 5% CO2; store frozen vials in the vapor phase of liquid nitrogen.
  • Human erythroleukemia cell line derived from bone marrow.
  • Suspension growth properties suitable for hematopoietic studies.
  • Grows in RPMI-1640 medium with 10% fetal bovine serum.
  • Incubate at 37°C in a 5% CO2 atmosphere.
  • Maintain cell density between 1 x 10^5 and 1 x 10^6 cells/mL.
  • Store frozen vials in vapor-phase liquid nitrogen.
Catalog No. NC3588088