Learn More
Apexbio Technology LLC Ticagrelor 274693-27-5 10mg
SDP
Supplier:  Apexbio Technology LLC B216610
Encompass_Preferred
Ticagrelor (CAS 274693-27-5) is an orally active reversible antagonist targeting the platelet P2Y12 receptor By competitively binding to P2Y12 ticagrelor inhibits ADP-mediated platelet aggregation resulting in dose-dependent suppression of platelet activation and thrombosis in vitro Unlike other P2Y12 inhibitors requiring metabolic activation ticagrelor acts without metabolic transformation though it is primarily metabolized by CYP3A4 and CYP2C19 enzymes This compound is frequently employed in biomedical research exploring platelet function thrombosis mechanisms and drug-drug interactions involving cytochrome P450 enzymes
Catalog No. 50-000-63703
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.