Learn More
Cayman Chemical Ticagrelor, 274693-27-5, 5 mg
SDP
Supplier:  Cayman Chemical 154255
Encompass_Preferred
Molecular Formula C23H28F2N6O4S, Purity ≥95%, Formula Weight 522.6, Synonyms: AR-C 126532XX, AZD 6140; A reversible purinergic P2Y12 receptor antagonist (Ki = 14 nM; IC50 = 1.8 M).
Catalog No. NC0944691
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.