Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TIE1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA527903
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA527903 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA527903 Supplier Invitrogen™ Supplier No. PA527903
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: HUVEC. Predicted reactivity: Mouse (97%), Rat (96%), Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Probable protein tyrosine-kinase transmembrane receptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TIE1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene TIE1
Gene Accession No. P35590
Gene Alias D430008P04Rik; JTK14; TIE; Tie1; Tie-1; tyrosine kinase receptor 1; tyrosine kinase with immunoglobulin and epidermal growth factor homology domains 1; tyrosine kinase with immunoglobulin like and EGF like domains 1; tyrosine kinase with immunoglobulin-like and EGF-like domains 1; tyrosine-protein kinase receptor Tie-1
Gene Symbols TIE1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human TIE1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7075
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.