Learn More
Apexbio Technology LLC Tie2 kinase inhibitor, 25mg. Cas: 948557-43-5 MFCD: MFCD18384977. Tie-2(Tie2 ) inhibitor
SDP
Supplier:  Apexbio Technology LLC A597925
Encompass_Preferred
IC50: A reversible and selective inhibitor of Tie2 with IC50 of 0.25 M, whose selectivity is 200-fold higher than that of p38. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4965
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.