Learn More
TIF1 Alpha/TRIM24 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A300816A
Description
The recommended shelf life for this product is 1 year from date of receipt.
The protein encoded by this gene mediates transcriptional control by interaction with the activation function 2 (AF2) region of several nuclear receptors, including the estrogen, retinoic acid, and vitamin D3 receptors. The protein localizes to nuclear bodies and is thought to associate with chromatin and heterochromatin-associated factors. The protein is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains - a RING, a B-box type 1 and a B-box type 2 - and a coiled-coil region. Two alternatively spliced transcript variants encoding different isoforms have been described for this gene.Specifications
| TIF1 Alpha/TRIM24 | |
| Polyclonal | |
| Unconjugated | |
| TRIM24 | |
| PTC6, RNF82, TF1A, TIF1, TIF1A, TIF1ALPHA, hTIF1, E3 ubiquitin-protein ligase TRIM24, RING finger protein 82, TIF1-alpha, transcription intermediary factor 1-alpha, transcriptional intermediary factor 1, tripartite motif-containing 24 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8805 | |
| 4° C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 1 mg/ml | |
| phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
| O15164 | |
| TRIM24 | |
| Between 1000 and C-term | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.