Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIG™ Tight-Head UN Rated 5 Gal. Carbon Steel Drum
SDP

Catalog No. 18366523 Shop All New Pig Corporation Products
Change view
Click to view available options
Capacity (English):
5 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
18-366-523 5 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-366-523 Supplier PIG™ Supplier No. DRM687LD
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tough, UN Rated drums that are made for hazardous materials.

  • UN Rated and tested to help ensure safe shipping of hazardous materials
  • Tight-head design is equipped with 2 in. NPT and 3/4 in. NPT bungs
  • Interior is coated with a corrosion inhibitor

Specifications

Product Type Lined Drum
Capacity (English) 5 gal.
Capacity (Metric) 18.9 L
Color Black
Material Steel
Height (English) 11.25 in.
Height (Metric) 29 cm
Diameter (English) Inner 14 in.
Diameter (English) Outer 14.5 in.
Diameter (Metric) Inner 35.56 cm
Diameter (Metric) Outer 37 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.