Learn More
AdipoGen TIGIT (human):Fc (human) (rec.) (50 µg)
SDP
Supplier:  AdipoGen AG40B0162C050
Encompass_Preferred
Protein. The extracellular domain of human TIGIT (aa 22-138) is fused at the C-terminus to the Fc portion of human IgG1. Source: HEK 293 cells. Endotoxin content: <0.01EU/µg purified protein (LAL test; Lonza). Lyophilized. Contains PBS. Purity: >95% (SDS-PAGE). TIGIT (T cell immunoreceptor with Ig and ITIM domains) is a novel member of the immunoglobulin super family (IgSF). TIGIT inhibits while CD226 activates the signaling through PVR and CD112. TIGIT and CD226 directly interact at the cell surface and this interaction impairs CD226 homodimerization and function. TIGIT is absent on naive T cells, but is expressed on activated Tregs and memory T cells, and on NK cells and NKT cells. TIGIT negatively regulates T cell activation and blockade of TIGIT exacerbates several immune diseases. Addition of TIGIT (human):Fc (human) to DCs triggers phosphorylation of PVR and elicits downstream signaling, which then inhibits T cell responses by producing IL-10.
Catalog No. 50-205-3255
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.