Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R3 Safety™ Tillman™ Truefit™ Goatskin Leather Performance Gloves

Catalog No. 19819959
Encompass
Change view
Click to view available options
Size:
Large
X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-819-959 Large
19-819-960 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-819-959 Supplier R3 Safety™ Supplier No. 1470L
This item is not returnable. View return policy

Top grain goatskin leather construction provides dexterity, while the spandex back ensures a secure fit.

  • Top grain goatskin leather construction provides dexterity
  • Spandex back ensures a secure fit
  • Leather fingertips for reinforcement in a high wear area
  • Smooth finger surface enhances grip capabilities
  • Double layered palm extends glove life
  • Elastic cuff with hook and loop closure
  • Reinforced thumb

Specifications

Back Material Spandex
Cuff Material Elastic
Color Black, Gray
Hand Specific
Material Top Grain Goatskin Leather
Product Type Mechanic's Gloves
Size Large
Finger Style Full
For Use With (Application) Warehouse Handling, Maintenance Work, and Powertool Handling
Glove Cuff Style Hook and Loop
Series 1470
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.