Learn More
Cayman Chemical 10 11-DIHDPA 500ug
SDP
Supplier:  Cayman Chemical 18174500 UG
Encompass_Preferred
A DHA metabolite that inhibits VEGF-induced angiogenesis in mice
Catalog No. 50-296-6743
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.