Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poltex™ Tilted Biohazard Bag Box
SDP

Catalog No. 22000796
Change view
Click to view available options
Height (Metric):
228 mm
381 mm
Length (Metric):
233 mm
386 mm
Width (Metric):
89 mm
152 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Height (Metric) Length (Metric) Width (Metric)
22-000-796 228 mm 233 mm 89 mm
22-000-809 381 mm 386 mm 152 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-000-796 Supplier Poltex™ Supplier No. BIOHTLT8
Add to Cart
Add to Cart

Benchtop biohazard bag holders allow for a cleanable mount for your biohazard bags.

  • Tilted toward the user for better ergonomics in BSC or other tight space
  • Comes in two sizes to fit either 8 x 12 in. or 14 x 19 in. biohazard bags
  • Constructed from super durable and cleanable PETG
  • Rubber bumpons on base for non-slip surface

Specifications

Autoclavable Not Autoclavable
Color Clear
Holds Bags
Material PETG
Positions 1
Shape Rectangular
Stackable No
Sterility Non-sterile
Length (Metric) 233 mm
Product Type Bag Rack
Shelves None
Height (Metric) 228 mm
Array 1 x 1
Width (Metric) 89 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.