Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ TIM-1/KIM-1/HAVCR Antibody Pair [HRP]
SDP

Catalog No. NB010268 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
15 Plates
5 Plates
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB010268 5 Plates
NB010267 15 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB010268 Supplier Novus Biologicals™ Supplier No. NBP2796145PLATES

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Rabbit Monoclonal Antibody Pair

specifically detects in Canine samples. It is validated for ELISA, Sandwich ELISA.

Specifications

Assay Range 125 pg/mL
Assay Sensitivity 125-8000 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type Antibody Pairs
Specificity This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Canine TIM-1/KIM-1/HAVCR.
Target Species Canine
For Use With (Application) Sandwich ELISA
Gene Alias HAVCR-1, HAVCRT cell immunoglobin domain and mucin domain protein 1, hepatitis A virus cellular receptor 1, Kidney injury molecule 1, KIM1, KIM-1, T-cell immunoglobulin and mucin domain-containing protein 1, TIM-1TIM, TIM1TIMD-1, TIMD1T-cell membrane protein 1
Gene ID (Entrez) 26762
Gene Symbol HAVCR1
Host Species Rabbit
Kit Contents
  • Standard: Each vial contains 500 ng of recombinant Canine TIM-1/KIM-1/HAVCR.
  • Capture Antibody: 1 mg/mL of rabbit TIM-1/KIM-1/HAVCR monoclonal antibody (in PBS, pH 7.4).
  • Detection Antibody: 1 mg/mL rabbit TIM-1/KIM-1/HAVCR monoclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % glycerol, pH 7.4).
  • Quantity 5 Plates
    Regulatory Status RUO
    Storage Requirements Storage is content dependent.
    Target TIM-1/KIM-1/HAVCR
    Technique 1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 1 μg/mL in detection antibody dilution buffer before use. 3. Standard: After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 8 ng/mL is recommended.
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.