Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TIM-3 (CD366) Mouse anti-Human, PE-CF594, Clone: 7D3, BD Horizon™

Catalog No. BDB565560 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB565560 100 Tests
BDB565561 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB565560 Supplier BD Biosciences Supplier No. 565560
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TIM-3 (CD366) Monoclonal antibody specifically detects TIM-3 (CD366) in Human samples. It is validated for Flow cytometry.

TIM-3 (CD366) Monoclonal antibody specifically detects TIM-3 (CD366) in Human samples. It is validated for Flow cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TIM-3 (CD366)
Applications Flow Cytometry
Classification Monoclonal
Clone 7D3
Conjugate PE-CF594
Description Vol. Per test: 5μL
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Gene Alias CD366; HAVCR2; TIM3; T cell immunoglobulin mucin-3; TIMD-3; KIM-3
Host Species Mouse
Immunogen Human TIM-3
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The antibody was conjugated with BD Horizon™ PE-CF594 under optimum conditions, and unconjugated antibody and free PE-CF594 were removed. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.