Learn More
TIM3 Mouse anti-Human, Mouse, Porcine, Rat, Clone: 4C4G3, Proteintech
Shop All Proteintech Group Inc Products

Description
TIM3 is a 280 aa membrane protein with a calculated molecular weight of 33 kDa, the higher molecular weights between 50 and 70 kDa detected by this monoclonal antibody probably represent glycosylated TIM3.
Specifications
Specifications
| Antigen | TIM3 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | 4C4G3 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | HAVCR2 |
| Gene Accession No. | P0C0K5, Q8TDQ0, Q8VIM0 |
| Gene Alias | CD366, HAVcr 2, HAVCR2, KIM 3, T cell membrane protein 3, Tim 3, TIM3, TIM-3, TIMD 3, TIMD3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.