Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TIMM10B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597081
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597081 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597081 Supplier Invitrogen™ Supplier No. PA597081
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MERQQQQQQQ LRNLRDFLLV YNRMTELCFQ RCVPSLHHRA LDAEEEACLH SCAGKLIHSN HRLMAAYVQL MPALVQRRIA DYEAASAVPG VAAEQPGVSP SGS; Positive Samples: 293T, HepG2, BT-474, HT-1080; Cellular Location: Mitochondrion inner membrane, Peripheral membrane protein.

FXC1, or TIMM10B, belongs to a family of evolutionarily conserved proteins that are organized in heterooligomeric complexes in the mitochondrial intermembrane space. These proteins mediate the import and insertion of hydrophobic membrane proteins into the mitochondrial inner membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TIMM10B
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.72 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Timm10b
Gene Accession No. Q9WV96, Q9Y5J6
Gene Alias fracture callus 1; fracture callus 1 homolog; fracture callus protein 1; fractured callus 1; fractured callus expressed transcript 1; FXC1; Mitochondrial import inner membrane translocase subunit Tim10 B; mitochondrial import inner membrane translocase subunit Tim9 B; small zinc finger-like protein; tim10b; Tim9b; TIMM10B; TIMM9B; translocase of inner mitochondrial membrane 10 homolog B (yeast); translocase of inner mitochondrial membrane 10B
Gene Symbols Timm10b
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-103 of human TIMM10B (NP_0363241).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14356, 26515
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.