Learn More
MyBioSource TIMM13 antibody
SDP
Supplier:  MyBioSource MBS96115240.2 ML
Encompass_Preferred
The TIMM13 antibody from MyBioSource is a rabbit polyclonal antibody raised against a recombinant protein of human TIMM13. This antibody detects endogenous levels of total TIMM13 protein. It can be used in Western blot, ELISA, and immunohistochemistry. The immunogen is a recombinant protein of human TIMM13. This antibody has been tested in human, mouse, and rat samples. The recommended dilution for Western blot is 1:500-1:1000, for ELISA it is 1:500-1:1000, and for IHC it is 1:50-1:200. This antibody is stored in PBS with 0.02% sodium azide and 50% glycerol at -20 C.
Catalog No. NC2874957
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.