Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TIMM21 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100224
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100224 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100224 Supplier Thermo Scientific Supplier No. PA5100224
Only null left
Add to Cart
Add to Cart

TIMM21 Polyclonal Antibody

TIMM21 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen TIMM21
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene TIMM21
Gene Accession No. Q8CCM6, Q9BVV7
Gene Alias 1700034H14Rik; 2700002I20Rik; C18orf55; HSPC154; Mitochondrial import inner membrane translocase subunit Tim21; Tim21; TIM21-like protein, mitochondrial; TIMM21; tranlocase of inner mitochondrial membrane 21; translocase of inner mitochondrial membrane 21; translocase of inner mitochondrial membrane 21 homolog
Gene Symbols TIMM21
Host Species Rabbit
Immunogen A synthesized peptide derived from human TIMM21(Accession Q9BVV7), corresponding to amino acid residues K49-Q99.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29090, 67105
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.