Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Timolol maleate

Catalog No. AC466510010
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC466510010 1 g
AC466512500 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC466510010 Supplier Thermo Scientific Chemicals Supplier No. 466510010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 26921-17-5 | C17H28N4O7S, C13H24N4O3S·C4H4O4 | 432.49 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26921-17-5
Molecular Formula C13H24N4O3S·C4H4O4

Specifications

Quantity 1 g
Solubility Solubility aq. soln.: soluble. Other solubilities: practically insoluble in ether, soluble in dmso, methanol, ethanol
Melting Point 202.0°C to 203.0°C
Formula Weight 432.49
InChI Key WLRMANUAADYWEA-NWASOUNVSA-N
SMILES OC(=O)\C=C/C(O)=O.CC(C)(C)NC[C@H](O)COC1=NSN=C1N1CCOCC1
IUPAC Name (2S)-1-(tert-butylamino)-3-{[4-(morpholin-4-yl)-1,2,5-thiadiazol-3-yl]oxy}propan-2-ol; (2Z)-but-2-enedioic acid
Molecular Weight (g/mol) 432.49
Chemical Name or Material Timolol maleate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Harmful if swallowed.

Suspected of damaging fertility or the unborn child.

Precautionary Statement GHS P Statement Obtain special instructions before use.

Wear protective gloves/protective clothing/eye protection/face protection.

Wash face, hands and any exposed skin thoroughly after handling.

IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

Call a POISON CENTER or doctor/physician if you feel unwell.

EINECSNumber 248-111-5

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.