Learn More
Invitrogen™ TIMP2 Polyclonal Antibody

Description
Does not cross react with other TIMP family members (TIMP1, TIMP3, TIMP4).
Specifications
Specifications
| Antigen | TIMP2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 15mM sodium azide; pH 7.4 |
| Gene | TIMP2 |
| Gene Accession No. | P16035 |
| Gene Alias | Collagenase inhibitor; CSC-21K; D11Bwg1104e; DDC8; etID32613.12; fa96h09; Metalloproteinase inhibitor 2; metalloproteinase inhibitor 2a; metalloproteinase inhibitor precursor; metalloproteinase inhibitor TIMP-2; metalloproteinase-2; similar to metalloproteinase inhibitor; testicular secretory protein Li 57; TIMP 2; TIMP metallopeptidase inhibitor 2; TIMP metallopeptidase inhibitor 2a; TIMP2; Timp-2; TIMP-2 protein; timp2a; tissue inhibitor of matrix metalloproteinase-2; tissue inhibitor of metalloproteinase 2; tissue inhibitor of metalloproteinase 2 precursor; tissue inhibitor of metalloproteinase 2a; tissue inhibitor of metalloproteinase-2; tissue inhibitor of metalloproteinase-2 precursor; tissue inhibitor of metalloproteinases 2; tissue inhibitor of mettaloproteinase 2; wu:fa96h09; zgc:136726 |
| Gene Symbols | TIMP2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.