Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tin, Foil 0.0125 mm Thickness, 300 X 300 mm 97.5%, Thermo Scientific™

Catalog No. AC420975000
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC420975000 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC420975000 Supplier Thermo Scientific Chemicals Supplier No. 420975000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-31-5
Molecular Formula Sn
Molecular Weight (g/mol) 118.71
MDL Number MFCD00133862
InChI Key ATJFFYVFTNAWJD-UHFFFAOYSA-N
Synonym powder, stannum, metallic, tin, elemental, wang, zinn, flake, tin, metal, silver matt powder, zinn german
PubChem CID 5352426
ChEBI CHEBI:32990
IUPAC Name tin
SMILES [Sn]

Specifications

Quantity 500 g
Form Foil
Linear Formula Sn
Fieser 01,1168; 02,414; 11,519; 12,486; 13,298; 17,333
Merck Index 15, 9603
Formula Weight 118.71
Percent Purity ≥97.5% (Sn)
Chemical Name or Material Tin, 97.5%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.