Learn More
Apexbio Technology LLC Tin Mesoporphyrin IX (chloride) 106344-20-1 25mg
SDP
Supplier:  Apexbio Technology LLC C560625
Encompass_Preferred
Tin Mesoporphyrin IX chloride is a small-molecule inhibitor targeting heme oxygenase (HO) It is designed to competitively inhibit HO an enzyme responsible for the oxidative degradation of heme to biliverdin carbon monoxide and ferrous iron thereby modulating heme metabolism and bilirubin formation Tin Mesoporphyrin IX chloride exerts its biological activity primarily through direct competition for the active sites of heme oxygenase In in vitro studies Tin Mesoporphyrin IX chloride demonstrates inhibition with reported inhibition constants (Ki) in the nanomolar range Based on these pharmacological properties Tin Mesoporphyrin IX chloride holds research potential in studies of heme metabolism bilirubin regulation hyperbilirubinemia modulation and exploration of heme oxygenase function in metabolic disorders
Catalog No. 50-000-88298
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.