Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Tin, Metal, granular and mossy
SCI_ED

Catalog No. S25609 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Reagent Grade - Mossy
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25609 100 g Reagent Grade - Mossy
S25609A 500 g Reagent Grade - Mossy
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. S25609
Add to Cart
Add to Cart
This item is not returnable. View return policy

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 7440-31-5
Molecular Formula Sn
Molecular Weight (g/mol) 118.71
MDL Number MFCD00133862
InChI Key ATJFFYVFTNAWJD-UHFFFAOYSA-N
Synonym powder, stannum, metallic, tin, elemental, wang, zinn, flake, tin, metal, silver matt powder, zinn german
PubChem CID 5352426
ChEBI CHEBI:32990
IUPAC Name tin
SMILES [Sn]

Specifications

Color Gray
Density 7.31g/mL
Linear Formula Sn
Quantity 100 g
Formula Weight 118.7
Grade Reagent
Packaging Reagent Grade - Mossy
Form Solid
Chemical Name or Material Tin
Safety handling Check Alert storage code Red
ShelfLife 5 years
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.