Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tin Plasma Grade Reference Standard, 1mL = 1mg Sn, 1000ppm Sn (Sn in 20% HCl), Ricca Chemical
SDP

Catalog No. 85014 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
120 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
85014 120 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 85014 Supplier Ricca Chemical Company Supplier No. 85014

Chemical Identifiers

CAS 7440-31-5 , 7647-01-0 , 7732-18-5
Molecular Formula Sn
Molecular Weight (g/mol) 118.71
MDL Number MFCD00133862
InChI Key ATJFFYVFTNAWJD-UHFFFAOYSA-N
PubChem CID 5352426
ChEBI CHEBI:32990
IUPAC Name tin
SMILES [Sn]

Specifications

Packaging Poly natural
Concentration or Composition Notes 1000ppm Sn (Sn in 20% HCl)
Equivalence 1 mL = 1 mg Sn
Physical Form Liquid
Color Colorless
Quantity 120 mL
Chemical Name or Material Tin Plasma Grade Reference Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.