Learn More
Selleck Chemical LLC AMI-1
Supplier: Selleck Chemical LLC S788425MG
AMI-1 is a potent and specific Histone Methyltransferase (HMT) inhibitor with IC50 of 3 0 M and 8 8 M for yeast Hmt1p and human PRMT1 respectively
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.