Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tingley™ Safetyflex Flame Resistant Coverall

Catalog No. 19056312 Shop All Tingley Rubber Co. Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-056-312 Medium
19-150-1307 X-Large
19-150-1306 Large
19-150-1308 2X-Large
19-150-1309 3X-Large
19-056-311 Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-056-312 Supplier Tingley™ Supplier No. J41008.MD
Add to Cart
Add to Cart
This item is not returnable. View return policy

Specifications

Size Medium
Material PVC, Polyester
Color Green
Neck Style Collared
Closure Type Storm Fly front
Seam Type Sealed
Resistance Flame, Acid, Chemicals, Caustics
Collar Type Standing
Certifications/Compliance ASTM D6413
For Use With (Application) Heavy exterior coating for excellent permeation performance
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.