Learn More
Apexbio Technology LLC Sulfanilamide 63-74-1 50g
SDP
Supplier:  Apexbio Technology LLC B204150000
Encompass_Preferred
Sulfanilamide is a competitive inhibitor targeting bacterial dihydropteroate synthetase (DHPS) an enzyme essential in microbial folate biosynthesis pathway By inhibiting DHPS the compound limits synthesis of dihydrofolate a precursor crucial for bacterial DNA replication and proliferation In enzymatic assays evaluating inhibition potency Sulfanilamide demonstrates an IC50 value of approximately 320 M Due to its defined mode of action Sulfanilamide is extensively utilized in microbiology and pharmacology research particularly for investigating bacterial metabolic pathways resistance mechanisms and screening potential antibacterial agents
Catalog No. 50-000-62291