Learn More
Sigma Organic Chemistry Tin(II) 2-ethylhexanoate | 1KG | 301-10-0 | MFCD00002676
Supplier: Sigma Organic Chemistry S32521KG
Tin(II) 2-ethylhexanoate, 1KG
About This Item:
Density: 1.251 g/mL at 25 C (lit.)
Refractive Index: n20/D 1.493 (lit.)
Storage: room temp
EINECS Number: 206-108-6
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.