Learn More
Strem, An Ascensus Company CAS 7646-78-8. 5g. Tin(IV) chloride anhydrous (99.99-Sn%) PURATREM.
SDP
Supplier:  Strem, An Ascensus Company 5050225G
Encompass_Preferred
CAS 7646-78-8. 5g. Tin(IV) chloride anhydrous (99.99-Sn%) PURATREM. . Molecular Weight 260.50. Molecular Formula SnCl4. Color/form colorless liq. Strem 50-5022. http//www.strem.com/catalog/v/50-5022/
Catalog No. 50-211-0848
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.