Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tinuvin P, Restek™
SDP

Catalog No. 06802060 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-802-060 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-802-060 Supplier Restek Corporation Supplier No. 31629

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2440-22-4
Molecular Formula C13H11N3O
Molecular Weight (g/mol) 225.251
InChI Key MCPKSFINULVDNX-UHFFFAOYSA-N
PubChem CID 17113
IUPAC Name 2-(benzotriazol-2-yl)-4-methylphenol
SMILES CC1=CC(=C(C=C1)O)N2N=C3C=CC=CC3=N2

Specifications

Packaging Ampule
CAS Max % 99.9%
Ignition Point 399° C
Odor Moderate
Vapor Density 2.07 (Air = 1)
Flash Point 54°C
Concentration 1.8 μg/mL in Isopropanol
UN Number UN 1219
Melting Point -90°C
Quantity 1 mL
Chemical Name or Material Isoprpropanal
Show More Show Less
Hazard Category Danger
Hazard Statement Highly flammable liquid and vapour. Causes serious eye irritation. May cause drowsiness or dizziness
Precautionary Statement Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Ground/bond container and receiving equipment.
Use explosion proof electrical/ventilation and lighting equipment.
Use only non - sparking tools.
Take precautionary measures against static discharge.
Avoid breathing dust/fume/gas/mist/vapours/spray.
Wash hands and skin thoroughly after handling
Use only outdoors or in a well - ventilated area.
Wear protective gloves/protective clothing/eye protection/face protection
EINECSNumber 200-746-91
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.