Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tiopronin 97.0+%, TCI America™
SDP

Catalog No. T261425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T261425G 25 g
T26145G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T261425G Supplier TCI America Supplier No. T261425G
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2-2-1953
Molecular Formula C5H9NO3S
Molecular Weight (g/mol) 163.19
MDL Number MFCD00004861
InChI Key YTGJWQPHMWSCST-UHFFFAOYNA-N
Synonym N-(2-Mercaptopropionyl)glycine
PubChem CID 5483
ChEBI CHEBI:32229
IUPAC Name 2-(2-sulfanylpropanoylamino)acetic acid
SMILES CC(C(=O)NCC(=O)O)S

Specifications

Melting Point 98°C
Color White-Yellow
Quantity 25 g
Formula Weight 163.19
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Tiopronin
EINECSNumber (2)-1207&(9)-1423
RTECSNumber MC0596500
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.