Learn More
Apexbio Technology LLC UK-5099 56396-35-1 50mg
SDP
Supplier:  Apexbio Technology LLC A389950
Encompass_Preferred
UK-5099 (CAS 56396-35-1) is a potent inhibitor targeting mitochondrial pyruvate carrier (MPC) a transporter facilitating pyruvate translocation across the mitochondrial inner membrane and serving vital functions in carbohydrate lipid and amino acid metabolism UK-5099 exhibits inhibition of pyruvate uptake with a reported Ki of 49 M and fully blocks transport activity at 1 mM Additionally IC50 values of approximately 50 nM have been observed for pyruvate-dependent oxygen consumption in rat liver and cardiac mitochondria Due to its inhibitory profile UK-5099 is frequently employed in metabolic studies to investigate mitochondrial pyruvate transport mechanisms and related biological pathways
Catalog No. 50-000-64003
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.