Learn More
CPC Scientific H-Ser-Leu-Ala-Leu-Ala-Asp-Asp-Ala-Ala-Phe-Arg-Glu-Arg-Ala-Arg-Leu-Leu-Ala-Ala-Leu-Glu-Arg-Arg-His-Trp-Leu-Asn-Ser-Tyr-Met-His-Lys-Leu-Leu-Val-Leu-Asp-Ala-Pro-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific PTHP017A
Encompass
This is a tuberoinfundibular neuropeptide and parathyroid hormone 2(PTH 2)-receptor agonist from hypothalmus. Synthetic TIP39 activates human and rat PTH2 receptors. ONE-LETTER SEQUENCE: SLALADDAAFRERARLLAALERRHWLNSYMHKLLVLDAPMOLECULAR FORMULA: C202H325N61O54S1MOLECULAR WEIGHT:4504.3STORAGE CONDITIONS: -20 5CRESEARCH AREA: OsteoporosisREFERENCES: Usdin, TB. et al. Nat. Am. 2, 941 (1999) Piserchio, A. et al. J. Biol. Chem. 275, 27284 (2000).
Catalog No. 50-211-0356