Learn More
Mascon Medical Mascon Medical™ Tip Racks, Suitable For 10μl Tips, 50EA/Case
SDP
Supplier:  Mascon Medical FSTIPRACK10
Encompass_Preferred
Our Mascon Medical™ Pipette Tip Empty Hinged Racks are the perfect solution for safe and secure storage of pipette tips. The front latch closes and locks to ensure samples are secure and are efficiently used for liquid handling, molecular biology, PCR and biotechnology.Features:•Meets ISO13485 quality management system standardSpecifications:•Volume: holds 96 10μl pipette tips•Qty. / Case: 50•Material: Polypropylene•Sterile: No•Temperature Range: -40℃~100℃•Packaging Format: 50 EA/Case
Catalog No. NC2458350
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.