Learn More
Andwin Scientific TIPONE 0.1-10 5L NATURAL ELO
SDP
Supplier:  Andwin Scientific 11103800
Encompass_Preferred
TipOne 0.1-10/20 ul XL natural ultra micro pipet tips in racks have a hydrophobic filter that are compsed of ultra high-density polyethylene. The filter traps aerosols and airborne contaminants in order to greatly reduce the risk of sample carry-over and cross contamination. This product contains improved 0.1-10/20 l XL ultra micro pipet tips fit most 0.1-2.5 l, 0.5-10 l, and 0.1-10 l pipettes. This product features improved graduation intervals: 1, 5, 10, and 20 l, holds up to 20 l, and has longer length to keep the tip collar outside of 1.5/2.0 ml tubes and most deep well plates, minimizing contamination. This product is certified free of RNase, DNase, DNA and pyrogens. This product comes in 10 racks of 96 tips (960 total tips).
Catalog No. 50-819-513
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.