Learn More
Andwin Scientific TIPONE 1-200 5L YELLOW BEVEL
SDP
Supplier:  Andwin Scientific 11111206
Encompass
TipOne 1-200 ul yellow,beveled pipet tips are available in stacks which saves space on the bench and on the storage shelf. The pipet tip's footprint is wider than most standard stacked tips to prevent overturning during use. Each tip layer locks into the one below it, preventing accidental separation. This product is certified free of RNase, DNase, DNA and pyrogens. This product comes in 5 layers of two 96 tip wafers (960 total tips).
Catalog No. 50-819-533
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.