Learn More
Andwin Scientific TIPONE 101-1000 5L BLUE GRAD
SDP
Supplier:  Andwin Scientific 11112721
Encompass
TipOne 101-1000 ul blue, graduated pipet tip refill wafers are graduated at 250, 500, and 1000 l. Refills save more space and plastic. The included Profile Collar improves fit and the included Profile Wall is thinner and clearer than before. This product uses snap-in wafers that drop into an empty tip rack. This product is certified free of RNase, DNase, DNA and pyrogens. This product comes in 10 racks of 96 tips (960 total tips).
Catalog No. 50-819-548
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.