Learn More
USA Scientific Inc TipOne RPT 100 uL prof Pt
Supplier:  USA Scientific Inc 11831540
Encompass
TipOne RPT 100 uL Profile Point ultra low retention filter tip refill starter pack. Includes one box with ten racks of filter tips and one box of ten filter tip refill cassettes (total 1920 tips). str.
Catalog No. 50-274-5480
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.