Learn More
Thomas Scientific MultiGrip™ 1000µL Tips, Blue Grips, Racked & Sterile, 576/pack
SDP
Supplier:  Thomas Scientific 1158T52
Encompass
MultiGrip™ pipet tips from Denville Scientific are the best tips for multichannel pipettors. This revolutionary tip is manufactured from two different resins which requires only 1/10 the sealing pressure of other leading pipet tips and reduces wear on pipettors. The soft seal offers a more universal fit, which standardizes most popular multichannel pipettors on one line of tips.
Catalog No. NC1601242