Learn More
Med Vet International 3M QualitatIVe Fit Test Apparatus Kit, Ft-30, Each
Supplier: Med Vet International 382499EA
This kit includes a collar, two nebulizers, a hood, a sensitivity solution with a bitter taste and a bitter-tasting fit test solution to allow for immediate testing This 3M™ fit test kit uses a solution of denatonium benzoate to emit a bitter taste that's easy to discern Use it with any particulate respirator or respirator with a particulate prefilter that uses gas or vapor This 3M™ qualitative fit test apparatus meets requirements for fit testing respirators under current OSHA rules to ensure safety The solution included in this kit makes it easy to test the fit of a respirator
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.