Learn More
Selleck Chemical LLC Tiragolumab-A2028-1MG
SDP
Supplier:  Selleck Chemical LLC A20281MG
Encompass_Preferred
Tiragolumab (RG6058 MTIG7192A) is a humanized IgG1/kappa monoclonal antibody that binds TIGIT to prevent its interaction with its ligand PVR (CD155)
Catalog No. 50-234-0769
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.