Learn More
Grainger Tire Coupling Hub WE10 Bore 1-5/8
Supplier:  Grainger 15E659
Encompass_Preferred
The TB Wood's Tire Coupling Hub is a 4-bolt angular tire coupling hub made of ductile iron with a black finish. It has a bore diameter of 1-1/4 inches and a load rating of 1750 ft-lbs static torque. This tire coupling hub meets SAE standard J706 and is designed for use with TB Wood's Dura-Flex couplings to connect rotating shafts. The coupling hub can accommodate shaft sizes from 1-1/8 to 1-5/16 inches using included sleeves and keys. It weighs approximately 3 lbs. This durable coupling hub is ideal for use in industrial power transmission applications.
Catalog No. 50-275-2124
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.