Learn More
Grainger Tire Coupling Hub WE2 Bore 7/8
Supplier:  Grainger 15E637
Encompass_Preferred
The TB Wood's Tire Coupling Element is a rubber coupling insert that connects shafts in high vibration applications. It has a 4-bolt design and is made of natural rubber for vibration dampening. This model has a 1-3/8 inch bore size, 1-1/2 inch outside diameter, and 3/4 inch width. It can be used with TB Wood's Dura-Flex couplings. The angular design allows up to 4 degrees of misalignment. The rubber material provides vibration isolation and protects equipment. This is an OEM replacement part for various industrial applications. The coupling absorbs shock and torque loads to reduce wear on connected shafts.
Catalog No. 50-275-2776
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.