Learn More
Grainger Tire Coupling Hub WE20 Bore 1-1/8
Supplier:  Grainger 15E662
Encompass_Preferred
The TB Wood's Tire Coupling Hub is a 3-angular coupling hub made of cast iron with a black finish. It has a bore size of 1-1/4 inches and an outside diameter of 3-1/4 inches. This coupling hub can be used to connect shafts in machinery transfers. It is rated for applications up to 1750 RPM and 140 degrees Fahrenheit. The overall width of the coupling hub is 1-3/4 inches. This is an industrial component commonly used for power transmission in industrial machinery.
Catalog No. 50-275-1602
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.