Learn More
Grainger Tire Coupling Hub WE3 Bore 1-3/8
Supplier:  Grainger 15E644
Encompass_Preferred
The TB Wood's Tire Coupling Hub is a 4-bolt angular tire coupling hub made of cast iron for industrial power transmission applications. It has a bore size of 1-1/4 inches, an outside diameter of 3-3/4 inches, a hub projection of 1-3/8 inches, and a grease fitting of 1/4-28 UNF 2A thread. This tire coupling hub conforms to AGMA class 7 and is rated for applications up to 1750 RPM.
Catalog No. 50-275-3364
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.