Learn More
Grainger Tire Coupling Hub WE3 Bore 7/8
Supplier:  Grainger 15E640
Encompass_Preferred
The TB Wood's Tire Coupling Hub is a 4-bolt angular tire coupling hub made of ductile iron with a black finish. It has a bore diameter of 1-1/4 inches and a load rating of 17,500 lbs dynamic. The overall dimensions are 5-1/2 inches x 4-3/4 inches x 2-3/4 inches. This tire coupling hub connects to wheels to build an in-line drive shaft system. It features a straight bore design and is designed for use with TB Wood's Dura-Flex couplings. The hub bolts directly to the wheel and couples to the drive shaft.
Catalog No. 50-275-3126
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.