Learn More
Grainger Tire Coupling Hub WE4 Bore 1-1/8
Supplier:  Grainger 15E646
Encompass_Preferred
The TB Wood's Tire Coupling Hub is a 4-bolt angular tire coupling hub made of ductile iron with a black finish. It has a bore diameter of 1-1/4 inches and a load rating of 1750 ft-lbs. This tire coupling hub connects to a tire coupling spider to make up a complete tire coupling assembly for use in industrial power transmission applications. The angular offset design allows adjustment of shaft misalignment. It is recommended for use with TB Wood's Dura-Flex tire couplings.
Catalog No. 50-274-9950
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.