Learn More
Grainger Tire Coupling Hub WE5 Bore 1
Supplier:  Grainger 15E651
Encompass_Preferred
The TB Wood's Tire Coupling Hub is a 4-bolt angular tire coupling hub made of ductile iron with a black finish. It has a bore diameter of 1-1/4 inches, a bolt circle diameter of 4-1/2 inches, and an overall length of 3-1/4 inches. This tire coupling hub is designed for use with V-belts and sheaves up to a 4-1/2 inch bolt circle diameter. It features an angular offset design and is recommended for industrial drivetrain applications requiring misalignment compensation. The maximum torque capacity is 475 ft-lbs.
Catalog No. 50-275-1236
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.