Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TISAB with CDTA (TISAB II), for Fluoride ISE, Reagents

Catalog No. 01336220
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-336-220 20 L
01-336-222 10 L
01-336-221 1 L
01-336-219 4 L
01-336-218 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 01-336-220 Supplier Reagents Holdings Llc Supplier No. C734021020F
Add to Cart
Add to Cart

TISAB with CDTA (TISAB II), for Fluoride ISE, Reagent, use for Fluoride analysis. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 64-19-7 , 64-19-7
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Total Ionic Strength Adjustment Buffer
IUPAC Name water
SMILES O

Specifications

Boiling Point 100°C
Color Colorless
For Use With (Application) Fluoride Analysis
For Use With (Equipment) Fluoride Meter
pH 5 to 5.5
Physical Form Liquid
Quantity 20 L
Solution Type Buffer
Packaging Cubitainer
Specific Gravity 1.0343
Sterility Non-sterile
Grade Reagent
Chemical Name or Material TISAB with CDTA (TISAB II), for Fluoride ISE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.