Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poltex Tissue Box Holder-4.5D

Holds and dispenses one large box of tissues, or two small boxes.

$44.55 - $55.70

Specifications

For Use With (Application) Healthcare
For Use With (Equipment) Tissue boxes
Material Acrylic
Product Type Tissue Box Holder
View More Specs

Alert:

Consumables shown for demonstration purposes only and are not included with the product.


Products 2
Catalog Number Mfr. No. Mounting Price Quantity & Availability  
Catalog Number Mfr. No. Mounting Price Quantity & Availability  
22-000-477
View Documents
Poltex
TISSUE4.5M
Magnet
Each for $55.70
Add to cart
 
22-000-478
View Documents
Poltex
TISSUE4.5W
Wall
Each for $44.55
Add to cart
 
Description

Description

  • Clean and shiny acrylic holder for tissue boxes
  • Comes in two sizes (2.2 in. and 5 in. deep)
  • Two mounting methods (wall and powerful magnet)
  • Constructed from durable 1/4 in. thick acrylic
  • See image for dimensions
  • Hardware not included for wall mount unit (-W). Customer must provide their own screw/anchor for their wall type.
Specifications

Specifications

Healthcare
Tissue boxes
Acrylic
Tissue Box Holder
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit